This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TCAM1 SIRNA Assays ›
Gene Symbol:
Tcam1
Gene Name:
testicular cell adhesion molecule 1
Gene Aliases:
4930570F09Rik, AI429105
Chromosome Location:
Chr.11: 106273079 - 106288158 on Build GRCm38
UniGene ID:
Mm.159608
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178414

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tcam1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029467.3 NP_083743.2 5 973
GenBank mRNA AK016263.1 - 5 843
AK076581.1 - 5 893
BC050746.1 - 897 6741794

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021673, XM_006247642, BC166495.

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
single organismal cell-cell adhesion
molecular_function
integrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-h5txz:80/100.66.79.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline