siRNA Details
| Gene Symbol: Lyrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285959.1 | NP_001272888.1 | 4 | 672 | ||
| NM_001285960.1 | NP_001272889.1 | 4 | 805 | |||
| NM_029610.3 | NP_083886.1 | 4 | 1056 | |||
| GenBank mRNA | AK009135.1 | - | 4 | 1059 | ||
| BC011339.1 | - | 4 | 846 | 3978191 | ||

Lyrm1
Genomic Map