siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199177.1 | NP_001186106.1 | 12, 13 | 1410 | ||
| NM_133752.3 | NP_598513.1 | 11, 12 | 1356 | |||
| GenBank mRNA | AB044138.1 | - | 11, 12 | 1348 | ||
| AK004715.1 | - | 12, 13 | 1366 | |||
| AK029157.1 | - | 11, 12 | 1314 | |||
| AK030835.1 | - | 12, 13 | 1364 | |||
| AK053861.1 | - | 12, 13 | 1425 | |||
| AK122311.1 | - | 11, 12 | 1284 | |||
| AK145620.1 | - | 11, 12 | 1308 | |||
| BC052704.1 | - | 11, 12 | 1298 | |||
| BC138665.1 | - | 11, 12 | 1237 | 8861686 | ||
| BC145959.1 | - | 11, 12 | 1237 | 40131262 | ||

Opa1
Genomic Map