siRNA Details
| Gene Symbol: 4930506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114312.1 | NP_001107784.1 | 17 | 3227 | ||
| NM_175172.4 | NP_780381.1 | 15 | 2913 | |||
| GenBank mRNA | AK082304.1 | - | 15 | 2914 | ||
| AK220405.1 | - | 17 | 3257 | |||
| BC030338.1 | - | 17 | 2799 | 2599101 | ||
| DV657531.1 | - | 1 | 618 | |||

4930506M07Rik
Genomic Map