siRNA Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175187.5 | NP_780396.2 | 4 | 390 | ||
| GenBank mRNA | AK013305.1 | - | 3 | 257 | ||
| AK051974.1 | - | 3 | 257 | |||
| AK088393.1 | - | 4 | 362 | |||
| BC037611.1 | - | 2 | 170 | |||
| BC051663.1 | - | 4 | 338 | 30020101 | ||
| BC060067.1 | - | 4 | 372 | |||
| BY239899.1 | - | 4 | 390 | |||

Tmem161b
Genomic Map