siRNA Details
| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006438.4 | NP_006429.2 | 1, 2 | 244 | ||
| GenBank mRNA | AB002631.1 | - | 1, 2 | 152 | ||
| AK291791.1 | - | 1, 2 | 244 | |||
| AY359038.1 | - | 1, 2 | 168 | |||
| BC103815.1 | - | 1, 2 | 188 | 40000432 | ||
| BC103816.1 | - | 1, 2 | 188 | 40000433 | ||

COLEC10
Genomic Map