This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930544G11RIK SIRNA Assays ›
Gene Symbol:
4930544G11Rik
Gene Name:
RIKEN cDNA 4930544G11 gene
Gene Aliases:
-
Chromosome Location:
Chr.6: 65952571 - 65954014 on Build GRCm38
UniGene ID:
Mm.389756
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 179925

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930544G11Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161773.1 NP_001155245.1 600
GenBank mRNA AK006363.1 - 1 560
AK016035.1 - 1 562
BC107181.1 - 1 509
BC107182.1 - 1 508
EU477372.1 - 1 388

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: CK603687.

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline