siRNA Details
Gene Symbol: 4930549C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026300.2 | NP_080576.1 | 2 | 600 | ||
GenBank mRNA | AK016074.1 | - | 2 | 601 | ||
BC048442.1 | - | 2 | 590 | 6704947 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930549C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026300.2 | NP_080576.1 | 2 | 600 | ||
GenBank mRNA | AK016074.1 | - | 2 | 601 | ||
BC048442.1 | - | 2 | 590 | 6704947 |