siRNA Details
| Gene Symbol: 1700129C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026461.2 | NP_080737.1 | 1 | 201 | ||
| GenBank mRNA | AK006084.1 | - | 1 | 203 | ||
| AK007313.1 | - | 1 | 201 | |||
| AK015075.1 | - | 1 | 188 | |||
| BC049552.1 | - | 1 | 201 | 6774189 | ||

1700129C05Rik
Genomic Map