This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MCOLN2 SIRNA Assays ›
Gene Symbol:
Mcoln2
Gene Name:
mucolipin 2
Gene Aliases:
3300002C04Rik, AI549968, C86638, TRPML2
Chromosome Location:
Chr.3: 146149796 - 146195514 on Build GRCm38
UniGene ID:
Mm.116862
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 180485

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mcoln2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005846.2 NP_001005846.1 2, 3 315
NM_026656.4 NP_080932.2 2, 3 343
GenBank mRNA AF503575.1 - 2, 3 332
AK014467.1 - 2, 3 246
AK019454.1 - 2, 3 212
AK150446.1 - 2, 3 311
AK150547.1 - 2, 3 311
AK152067.1 - 2, 3 311
AK152943.1 - 2, 3 311
AK153290.1 - 2, 3 311
AK159172.1 - 2, 3 343
AY083532.1 - 1, 2 161
BC029847.1 - 2, 3 192 3967456
CJ134616.1 - 2, 3 315

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039005, BC099787.

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s)

transport
ion transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline