This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM114A1 SIRNA Assays ›
Gene Symbol:
Fam114a1
Gene Name:
family with sequence similarity 114, member A1
Gene Aliases:
1190001N04Rik, 9130005N14Rik, Noxp20
Chromosome Location:
Chr.5: 64970075 - 65041901 on Build GRCm38
UniGene ID:
Mm.258545
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 180499

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam114a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026667.3 NP_080943.3 6 831
GenBank mRNA AK007950.1 - 6 789
AK014725.1 - 6 801
AK017336.1 - 6 791
AK020262.1 - 6 792
AK030360.1 - 6 832
AM162548.1 - 6 835
BC027235.1 - 6 794 3603083
BC110359.1 - 6 832 5325099

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770177, XM_237514, XM_006250991, XM_008770176.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline