This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FASTKD3 SIRNA Assays ›
Gene Symbol:
Fastkd3
Gene Name:
FAST kinase domains 3
Gene Aliases:
2310010B21Rik, AW048587
Chromosome Location:
Chr.13: 68582163 - 68592338 on Build GRCm38
UniGene ID:
Mm.23670
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 180798

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fastkd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027123.4 NP_081399.3 2 311
GenBank mRNA AK031142.1 - 2 298
AK032530.1 - 2 296
AK033189.1 - 2 312
AK036778.1 - 2 297
AK148902.1 - 2 301
BC042506.1 - 2 288 4023412
BC060688.1 - 2 531 6409274

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular respiration
protein kinase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9wwlr:80/100.66.77.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline