This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ACSS1 SIRNA Assays ›
Gene Symbol:
Acss1
Gene Name:
acyl-CoA synthetase short-chain family member 1
Gene Aliases:
1110032O15Rik, AI788978, Acas2, Acas2l, AceCS2
Chromosome Location:
Chr.2: 150618111 - 150668240 on Build GRCm38
UniGene ID:
Mm.7044
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 181097

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acss1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080575.2 NP_542142.1 9, 10 1482
GenBank mRNA AB046742.1 - 9, 10 1471
AK088244.1 - 9, 10 1461
AK146568.1 - 9, 10 1483
AK158119.1 - 9, 10 1477
AK172412.1 - 9, 10 1468
AK172470.1 - 9, 10 1457
AK173275.1 - 9, 10 1443
BC030930.1 - 5, 6 656 4911679
BC092278.1 - 9, 10 1511 3710505

Back To Top

More Information


Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA biosynthetic process
metabolic process
acetate biosynthetic process
acetyl-CoA biosynthetic process from acetate
propionate biosynthetic process
nucleotide binding
catalytic activity
acetate-CoA ligase activity
ATP binding
AMP binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4zngx:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline