siRNA Details
| Gene Symbol: 2900011O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144518.3 | NP_653101.1 | 6 | 885 | ||
| GenBank mRNA | AK035868.1 | - | 6 | 862 | ||
| AK049997.1 | - | 6 | 917 | |||
| AK076136.1 | - | 6 | 904 | |||
| AK136859.1 | - | 6 | 885 | |||
| AK139470.1 | - | 6 | 897 | |||
| BC022741.1 | - | 6 | 864 | 4511454 | ||

2900011O08Rik
Genomic Map