This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VWA9 SIRNA Assays ›
Gene Symbol:
Vwa9
Gene Name:
von Willebrand factor A domain containing 9
Gene Aliases:
2010321M09Rik
Chromosome Location:
Chr.9: 64960832 - 64986981 on Build GRCm38
UniGene ID:
Mm.272616
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 181400

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vwa9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077631.2 NP_001071099.1 11 1665
NM_175153.4 NP_780362.2 11 1886
GenBank mRNA AK032095.1 - 11 1880
AK035654.1 - 11 1593
AK168222.1 - 10 1568
AK169772.1 - 11 1625

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001007663, XM_008766239, XM_008766238, BC082006, FQ231531.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0