siRNA Details
| Gene Symbol: Sav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022028.2 | NP_071311.1 | 5 | 1415 | ||
| GenBank mRNA | AJ292968.1 | - | 5 | 1415 | ||
| AK006655.1 | - | 4 | 852 | |||
| AK034983.1 | - | 3 | 2288 | |||
| BC019377.1 | - | 5 | 1348 | 4021056 | ||

Sav1
Genomic Map