siRNA Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023232.3 | NP_075721.3 | 3, 4 | 361 | ||
| GenBank mRNA | AF203914.1 | - | 3, 4 | 327 | ||
| AK002887.1 | - | 3, 4 | 331 | |||
| AK012760.1 | - | 3, 4 | 359 | |||
| AK077198.1 | - | 3, 4 | 342 | |||
| AK082903.1 | - | 3, 4 | 331 | |||
| AK133194.1 | - | 341 | ||||
| AK169573.1 | - | 3, 4 | 331 | |||
| BC024780.1 | - | 3, 4 | 333 | 5365266 | ||
| BC054109.1 | - | 3, 4 | 355 | 30013833 | ||

Diablo
Genomic Map