siRNA Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024194.6 | NP_077156.2 | 3 | 419 | ||
| GenBank mRNA | AK011726.1 | - | 3 | 400 | ||
| AK017814.1 | - | 3 | 388 | |||
| AK034978.1 | - | 399 | ||||
| AK148748.1 | - | 3 | 378 | |||
| AK163288.1 | - | 3 | 387 | |||

Lrrc40
Genomic Map