siRNA Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025388.2 | NP_079664.1 | 7 | 945 | ||
| GenBank mRNA | AK003070.1 | - | 6 | 847 | ||
| AK003897.1 | - | 7 | 946 | |||
| BC021936.1 | - | 7 | 906 | 5124904 | ||
| BC055109.1 | - | 2 | 1736 | |||
| Y17677.1 | - | 1 | 333 | |||

Ufc1
Genomic Map