This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700029H14RIK SIRNA Assays ›
Gene Symbol:
1700029H14Rik
Gene Name:
RIKEN cDNA 1700029H14 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 13550722 - 13562554 on Build GRCm38
UniGene ID:
Mm.46136
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183107

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700029H14Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080781.2 NP_001074250.1 2 334
NM_025601.3 NP_079877.1 2 334
GenBank mRNA AK006340.1 - 2 279
AK006493.1 - 2 277
AK016674.1 - 2 279
BC049575.1 - 2 300 6773883
CA464636.1 - 2 324

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fr92q:80/100.66.78.240:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0