This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EDDM3A SIRNA Assays ›
Gene Symbol:
EDDM3A
Gene Name:
epididymal protein 3A
Gene Aliases:
EP3A, FAM12A, HE3-ALPHA, HE3A, HE3ALPHA, RAM1
Chromosome Location:
Chr.14: 20735946 - 20748380 on Build GRCh38
UniGene ID:
Hs.304757
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 18351

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EDDM3A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006683.4 NP_006674.2 2 506
XM_017020934.1 XP_016876423.1 2 523
GenBank mRNA BC069407.1 - 2 477 7262138
BC096698.1 - 2 496 40014618
BC096731.1 - 2 496 40014622
BC098164.1 - 1 496 40014621
BC098356.1 - 1 496 40014616
BC106711.1 - 1 460 40009240
BC106712.1 - 1 460 40009242
X76383.2 - 2 506

Back To Top

More Information


Gene Ontology Categories:

Function(s)

sperm displacement

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-zzdf4:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline