siRNA Details
| Gene Symbol: Zcchc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035509.1 | NP_001030586.1 | 4 | 955 | ||
| NM_001035510.1 | NP_001030587.1 | 2 | 1426 | |||
| NM_025893.2 | NP_080169.2 | 4 | 903 | |||
| GenBank mRNA | AK005317.1 | - | 4 | 881 | ||
| AK075607.1 | - | 1 | 507 | |||
| AK134856.1 | - | 1284 | ||||
| AY650116.1 | - | 1 | 385 | |||
| BC017627.1 | - | 4 | 816 | 4505033 | ||

Zcchc18
Genomic Map