siRNA Details
| Gene Symbol: Syap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025932.2 | NP_080208.2 | 5 | 758 | ||
| GenBank mRNA | AK008390.1 | - | 5 | 727 | ||
| AK014893.1 | - | 5 | 744 | |||
| AK051251.1 | - | 5 | 715 | |||
| AK147055.1 | - | 5 | 694 | |||
| AK154843.1 | - | 5 | 715 | |||
| BC021373.1 | - | 5 | 688 | 5039534 | ||

Syap1
Genomic Map