This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TEX40 SIRNA Assays ›
Gene Symbol:
Tex40
Gene Name:
testis expressed 40
Gene Aliases:
1700019N12Rik, A430107B04Rik, MLZ-622
Chromosome Location:
Chr.19: 6922426 - 6925380 on Build GRCm38
UniGene ID:
Mm.251306
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183729

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tex40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039494.2 NP_001034583.1 2 232
GenBank mRNA AK005644.1 - 2 211
AK006034.1 - 2 170
AK006134.1 - 2 235
AK007089.1 - 2 208
BC048452.1 - 2 169
BC061075.1 - 2 174
BC172038.1 - 1 389
BC172041.1 - 1 389
BC172042.1 - 1 389

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760207, XM_008760206, XM_008760205.

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
biological_process
cell differentiation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline