This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IL1RAPL2 SIRNA Assays ›
Gene Symbol:
Il1rapl2
Gene Name:
interleukin 1 receptor accessory protein-like 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 137571549 - 138846955 on Build GRCm38
UniGene ID:
Mm.328321
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183850

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il1rapl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030688.1 NP_109613.1 4 562
GenBank mRNA AF284437.1 - 4 562
AJ277831.1 - 1320
AK039155.1 - 1087
AK045976.1 - 1351
BC119274.1 - 4 505 8734023
BC120555.1 - 4 505 40129478

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001166342, XM_008773406.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
interleukin-1, Type II, blocking receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline