This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM13A SIRNA Assays ›
Gene Symbol:
Fam13a
Gene Name:
family with sequence similarity 13, member A
Gene Aliases:
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location:
Chr.6: 58933536 - 59024502 on Build GRCm38
UniGene ID:
Mm.28813
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183938

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam13a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153574.2 NP_705802.1 5 551
GenBank mRNA AJ510136.1 - 5 537
AK052421.1 - 5 492
AK054311.1 - 5 538
BC005554.1 - 5 456
BC054053.1 - 5 525
BC060978.1 - 5 488
BC080861.1 - 5 551 30357292

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline