siRNA Details
| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018828.2 | NP_061298.1 | 2, 3 | 561 | ||
| GenBank mRNA | AK012167.1 | - | 2, 3 | 374 | ||
| AK014018.1 | - | 2, 3 | 378 | |||
| AK084018.1 | - | 2, 3 | 353 | |||
| AK129263.1 | - | 2, 3 | 458 | |||
| AK164543.1 | - | 2, 3 | 332 | |||
| U40750.2 | - | 2, 3 | 561 | |||

Fnbp4
Genomic Map