This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RCAN1 SIRNA Assays ›
Gene Symbol:
Rcan1
Gene Name:
regulator of calcineurin 1
Gene Aliases:
Adapt78, CALP1L, CSP1, DSC1, Dscr1, MCIP1, RCN1
Chromosome Location:
Chr.16: 92391951 - 92466169 on Build GRCm38
UniGene ID:
Mm.265744
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 185188

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rcan1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081549.2 NP_001075018.1 2 415
NM_019466.4 NP_062339.2 2 261
GenBank mRNA AF237789.1 - 2 165
AF237790.1 - 2 165
AF260717.1 - 2 179
AF263239.1 - 2 195
AF263240.1 - 2 278
AF282255.1 - 2 172
AK010696.1 - 132
AK077283.1 - 2 261
AK146764.1 - 2 390
AY325904.1 - 2 371
BC013551.1 - 2 241 4236038

Back To Top

More Information


Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

response to ischemia
response to stress
response to oxidative stress
short-term memory
calcium-mediated signaling
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
regulation of phosphoprotein phosphatase activity
skeletal muscle fiber development
regulation of catalytic activity
regulation of calcineurin-NFAT signaling cascade
protein binding
calcium-dependent protein serine/threonine phosphatase regulator activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4zngx:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline