This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VPS25 SIRNA Assays ›
Gene Symbol:
Vps25
Gene Name:
vacuolar protein sorting 25 (yeast)
Gene Aliases:
1110020N13Rik, AW107467, D11Wsu68e
Chromosome Location:
Chr.11: 101253611 - 101259547 on Build GRCm38
UniGene ID:
Mm.301020
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 186170

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284411.1 NP_001271340.1 4 338
NM_001284412.1 NP_001271341.1 4 314
NM_001284414.1 NP_001271343.1 4 314
NM_026776.4 NP_081052.2 4 314
GenBank mRNA AK002500.1 - 4 276
AK003715.2 - 4 286
AK003877.1 - 4 272
AK082933.1 - 4 293
AK145811.1 - 4 285
AK150822.1 - 4 296
AK162230.1 - 4 287
AK166008.1 - 4 293
AK190530.1 - 2 68
AK214794.1 - 2 68
BC029181.1 - 4 278 4240500

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_032353, AB014763, AK312092, BC006282.
This siRNA also targets the following transcripts in Rat: NM_001173451, FQ229682, FQ220187, FQ233075, FQ213477.

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transport
protein transport
protein targeting to vacuole involved in ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
structural molecule activity
protein homodimerization activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-r4kfh:80/100.66.79.35:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0