This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LARP7 SIRNA Assays ›
Gene Symbol:
Larp7
Gene Name:
La ribonucleoprotein domain family, member 7
Gene Aliases:
C330027G06Rik, D3Wsu161e
Chromosome Location:
Chr.3: 127536714 - 127553349 on Build GRCm38
UniGene ID:
Mm.291032
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 186360

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Larp7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138593.2 NP_613059.2 12 1763
GenBank mRNA AK012726.1 - 1 65
BC029178.1 - 4 436
BC132266.1 - 4 434
BC132268.1 - 4 434
BM946683.1 - 6 631

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
RNA processing
negative regulation of chromatin binding
germ cell proliferation
positive regulation of G1/S transition of mitotic cell cycle
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline