siRNA Details
| Gene Symbol: Skp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011543.4 | NP_035673.3 | 6 | 655 | ||
| GenBank mRNA | AF083214.1 | - | 6 | 579 | ||
| AK002983.1 | - | 6 | 626 | |||
| AK010628.1 | - | 6 | 641 | |||
| AK012498.1 | - | 6 | 614 | |||
| AK014245.1 | - | 6 | 614 | |||
| AK027909.1 | - | 6 | 613 | |||
| AK078330.1 | - | 6 | 641 | |||
| AK088339.1 | - | 6 | 614 | |||
| AK152609.1 | - | 6 | 613 | |||
| AK160198.1 | - | 6 | 615 | |||
| AK166673.1 | - | 6 | 614 | |||
| BC002115.1 | - | 6 | 598 | 3490159 | ||
| Z47088.1 | - | 6 | 604 | |||

Skp1a
Genomic Map