siRNA Details
| Gene Symbol: Tnfrsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077509.1 | NP_001070977.1 | 7, 8 | 793 | ||
| NM_011612.2 | NP_035742.1 | 7, 8 | 812 | |||
| GenBank mRNA | AK019885.1 | - | 7, 8 | 779 | ||
| AK154625.1 | - | 7, 8 | 796 | |||
| DQ832278.1 | - | 6, 7 | 698 | |||

Tnfrsf9
Genomic Map