siRNA Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162947.1 | NP_001156419.1 | 2, 3 | 254 | ||
| NM_011848.4 | NP_035978.2 | 3, 4 | 312 | |||
| GenBank mRNA | AF093416.1 | - | 2, 3 | 259 | ||
| AF099066.1 | - | 3, 4 | 281 | |||
| BC005411.1 | - | 3, 4 | 289 | 3590866 | ||
| BU058938.1 | - | 3, 4 | 312 | |||

Nek3
Genomic Map