This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRPC7 SIRNA Assays ›
Gene Symbol:
Trpc7
Gene Name:
transient receptor potential cation channel, subfamily C, member 7
Gene Aliases:
TRP7, Trrp8
Chromosome Location:
Chr.13: 56773098 - 56895974 on Build GRCm38
UniGene ID:
Mm.62409
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 188272

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trpc7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF139923.1 - 3 1028
AK135153.1 - 3 1217
AK137418.1 - 3 1229
BC117995.1 - 3 1028 40096022

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel transporter

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline