siRNA Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013736.4 | NP_038764.2 | 3 | 209 | ||
| GenBank mRNA | AB025015.1 | - | 3 | 193 | ||
| AK036592.1 | - | 3 | 210 | |||
| AK145994.1 | - | 3 | 210 | |||
| AK147088.1 | - | 3 | 204 | |||
| BC049885.1 | - | 3 | 205 | 5387774 | ||

Tceb3
Genomic Map