siRNA Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013736.4 | NP_038764.2 | 4 | 446 | ||
| GenBank mRNA | AK036592.1 | - | 4 | 447 | ||
| AK145994.1 | - | 4 | 447 | |||
| AK147088.1 | - | 4 | 441 | |||
| BC049885.1 | - | 4 | 442 | 5387774 | ||

Tceb3
Genomic Map