This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DEF6 SIRNA Assays ›
Gene Symbol:
Def6
Gene Name:
differentially expressed in FDCP 6
Gene Aliases:
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6
Chromosome Location:
Chr.17: 28207778 - 28228608 on Build GRCm38
UniGene ID:
Mm.204731
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 188913

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Def6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027185.3 NP_081461.2 3 323
GenBank mRNA AF329497.1 - 3 258
AK038050.1 - 3 294
AK088460.1 - 3 272
AY241695.1 - 3 258
BC120500.1 - 3 323 40129423
BC131953.1 - 3 323 40130230
X96705.1 - 3 215

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline