siRNA Details
| Gene Symbol: Cyp2c7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017158.2 | NP_058854.1 | 1, 2 | 181 | ||
| XM_008759609.1 | XP_008757831.1 | 1, 2 | 303 | |||
| GenBank mRNA | BC097939.1 | - | 1, 2 | 164 | 7368637 | |
| BC127503.1 | - | 1, 2 | 220 | 7371672 | ||
| FQ218330.1 | - | 1, 2 | 181 | |||
| M18335.1 | - | 1, 2 | 182 | |||
| M31031.1 | - | 1, 2 | 139 | |||

Cyp2c7
Genomic Map