siRNA Details
| Gene Symbol: Nrxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021767.2 | NP_068535.2 | 1780 | |||
| GenBank mRNA | M96374.1 | - | 1780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrxn1| Gene Symbol: Nrxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021767.2 | NP_068535.2 | 1780 | |||
| GenBank mRNA | M96374.1 | - | 1780 | |||