This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ACVR1 SIRNA Assays ›
Gene Symbol:
Acvr1
Gene Name:
activin A receptor, type I
Gene Aliases:
-
Chromosome Location:
Chr.3: 44432476 - 44551733 on Build Rnor_6.0
UniGene ID:
Rn.87899
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 189645

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acvr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024486.1 NP_077812.1 9 1439
XM_006234214.2 XP_006234276.1 11 3044
XM_008761862.1 XP_008760084.1 12 3101
XM_008761863.1 XP_008760085.1 12 5345
XM_008761864.1 XP_008760086.1 12 5091
XM_008761865.1 XP_008760087.1 12 5134
XM_008761866.1 XP_008760088.1 12 1811
XM_008761867.1 XP_008760089.1 9 1355
GenBank mRNA BC167754.1 - 10 1610 9089038
L19341.1 - 9 1439

Back To Top

More Information


Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
patterning of blood vessels
urogenital system development
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
neural crest cell migration
acute inflammatory response
embryonic heart tube morphogenesis
mitral valve morphogenesis
atrial septum primum morphogenesis
protein phosphorylation
transforming growth factor beta receptor signaling pathway
germ cell development
determination of left/right symmetry
gastrulation
mesoderm development
heart development
negative regulation of signal transduction
positive regulation of pathway-restricted SMAD protein phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
regulation of ossification
positive regulation of bone mineralization
BMP signaling pathway
activin receptor signaling pathway
negative regulation of activin receptor signaling pathway
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of skeletal muscle tissue development
smooth muscle cell differentiation
pharyngeal system development
pathway-restricted SMAD protein phosphorylation
cardiac muscle cell fate commitment
endocardial cushion cell fate commitment
cellular response to glucocorticoid stimulus
positive regulation of determination of dorsal identity
negative regulation of extrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
ATP binding
activin receptor activity, type I
growth factor binding
protein homodimerization activity
SMAD binding
metal ion binding
activin binding
transforming growth factor beta binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline