siRNA Details
| Gene Symbol: C3ar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032060.1 | NP_114449.1 | 2 | 902 | ||
| XM_006237314.2 | XP_006237376.1 | 2 | 1139 | |||
| GenBank mRNA | U86379.1 | - | 2 | 902 | ||
More Information
Set Membership: |
|

C3ar1
Genomic Map