siRNA Details
| Gene Symbol: Cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053593.2 | NP_446045.1 | 3 | 462 | ||
| GenBank mRNA | BC070956.1 | - | 3 | 462 | 7104124 | |
| FQ214013.1 | - | 3 | 478 | |||
| FQ219275.1 | - | 3 | 438 | |||
| FQ234663.1 | - | 2 | 91 | |||
| L11007.1 | - | 3 | 394 | |||

Cdk4
Genomic Map