siRNA Details
| Gene Symbol: Blvra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053850.1 | NP_446302.1 | 3 | 246 | ||
| XM_006234894.2 | XP_006234956.1 | 4 | 386 | |||
| XM_006234895.2 | XP_006234957.1 | 4 | 371 | |||
| GenBank mRNA | BC078766.1 | - | 3 | 258 | 7126157 | |
| FQ213399.1 | - | 4 | 325 | |||
| FQ230935.1 | - | 3 | 205 | |||
| FQ231949.1 | - | 4 | 327 | |||
| M81681.1 | - | 3 | 246 | |||

Blvra
Genomic Map