This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF146 SIRNA Assays ›
Gene Symbol:
ZNF146
Gene Name:
zinc finger protein 146
Gene Aliases:
OZF
Chromosome Location:
Chr.19: 36214602 - 36238774 on Build GRCh38
UniGene ID:
Hs.643436
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 19087

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF146
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099638.1 NP_001093108.1 3 1709
NM_001099639.1 NP_001093109.1 3 1668
NM_007145.2 NP_009076.2 4 1783
XM_005259214.2 XP_005259271.1 4 1608
XM_017027244.1 XP_016882733.1 3 1234
XM_017027245.1 XP_016882734.1 4 1239
XM_017027246.1 XP_016882735.1 3 1138
XM_017027247.1 XP_016882736.1 3 1200
GenBank mRNA AK311486.1 - 1 253
AK313460.1 - 1 721
BC065568.1 - 4 1252
BC110500.1 - 1 1009 40028224
BC110501.1 - 1 1009 40028226
DA645504.1 - 1 310
X70394.1 - 2 1116

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
heparin binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0