This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM107A SIRNA Assays ›
Gene Symbol:
FAM107A
Gene Name:
family with sequence similarity 107 member A
Gene Aliases:
DRR1, TU3A
Chromosome Location:
Chr.3: 58564112 - 58627610 on Build GRCh38
UniGene ID:
Hs.506357
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 19106

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM107A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076778.2 NP_001070246.1 3, 4 786
NM_001282713.1 NP_001269642.1 3, 4 586
NM_001282714.1 NP_001269643.1 4, 5 1034
NM_007177.3 NP_009108.1 4, 5 884
GenBank mRNA AB023811.1 - 3, 4 366
AF035283.1 - 3, 4 786
AF089853.1 - 3, 4 877
AF089854.1 - 4, 5 975
AK054720.1 - 4, 5 1034
AK128791.1 - 3, 4 413
AK298001.1 - 3, 4 541
AK316450.1 - 3, 4 441
AL050264.1 - 3, 4 399
BC010561.1 - 4, 5 485 4152118
BX645008.1 - 3, 4 596
CR457133.1 - 2, 3 325
EU569833.1 - 3, 4 441

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline