This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESM1 SIRNA Assays ›
Gene Symbol:
ESM1
Gene Name:
endothelial cell specific molecule 1
Gene Aliases:
endocan
Chromosome Location:
Chr.5: 54977867 - 54985586 on Build GRCh38
UniGene ID:
Hs.129944
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 19124

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ESM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135604.1 NP_001129076.1 1 322
NM_007036.4 NP_008967.1 1 322
GenBank mRNA AJ973643.1 - 1 269
AK300634.1 - 1 329
AK311817.1 - 1 329
BC011989.1 - 1 312 3882426
BM423977.1 - 1 399
X89426.1 - 1 308

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
regulation of cell growth
sprouting angiogenesis
positive regulation of cell proliferation
positive regulation of hepatocyte growth factor receptor signaling pathway
hepatocyte growth factor receptor binding
integrin binding
insulin-like growth factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline