This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AGO2 SIRNA Assays ›
Gene Symbol:
Ago2
Gene Name:
argonaute RISC catalytic component 2
Gene Aliases:
Eif2c2
Chromosome Location:
Chr.7: 114339607 - 114377277 on Build Rnor_6.0
UniGene ID:
Rn.234328
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 191905

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ago2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021597.1 NP_067608.1 2 260
GenBank mRNA AF195534.1 - 2 260

Back To Top

More Information


Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA cleavage
biological_process
post-embryonic development
miRNA metabolic process
positive regulation of gene expression
cell differentiation
gene silencing by RNA
pre-miRNA processing
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
negative regulation of translational initiation
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
RNA phosphodiester bond hydrolysis, endonucleolytic
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
RNA 7-methylguanosine cap binding
molecular_function
RNA binding
mRNA binding
endoribonuclease activity
siRNA binding
miRNA binding
poly(A) RNA binding
metal ion binding
endoribonuclease activity, cleaving siRNA-paired mRNA
pre-miRNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline