siRNA Details
| Gene Symbol: Nrxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053846.1 | NP_446298.1 | 22 | 5524 | ||
| XM_008760064.1 | XP_008758286.1 | 7 | 2619 | |||
| GenBank mRNA | M96376.1 | - | 22 | 5524 | ||
| M96377.1 | - | 7 | 2599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrxn2| Gene Symbol: Nrxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053846.1 | NP_446298.1 | 22 | 5524 | ||
| XM_008760064.1 | XP_008758286.1 | 7 | 2619 | |||
| GenBank mRNA | M96376.1 | - | 22 | 5524 | ||
| M96377.1 | - | 7 | 2599 | |||