This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MARF1 SIRNA Assays ›
Gene Symbol:
Marf1 LOC103689943
Gene Name:
meiosis arrest female 1
meiosis arrest female protein 1-like
Gene Aliases:
Lkap
-
Chromosome Location:
Chr.10: 908806 - 953481 on Build Rnor_6.0
Chromosome Location:
Chr.10: 28269 - 72912 on Build Rnor_6.0
UniGene ID:
Rn.27366
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 193305

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Marf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133421.2 NP_596912.1 3 360
XM_003750779.3 XP_003750827.1 3 1407
XM_006245854.2 XP_006245916.1 3 334
XM_006245855.2 XP_006245917.1 3 186
XM_006245856.2 XP_006245918.1 3 334
XM_008767442.1 XP_008765664.1 2 841
GenBank mRNA AB012133.1 - 3 360
Gene Symbol: LOC103689943
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245686.2 XP_006245748.1 3 1408
XM_006245687.2 XP_006245749.1 3 334
XM_006245688.1 XP_006245750.1 3 186
XM_006245689.2 XP_006245751.1 3 334
XM_008767447.1 XP_008765669.1 3 334
XM_008767448.1 XP_008765670.1 2 841

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081154, AK220341, AK035704.

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
female meiotic division
regulation of gene expression
negative regulation of phosphatase activity
oogenesis
nucleotide binding
RNA binding
nucleic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-mnvx7:80/100.66.78.106:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0