siRNA Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138531.2 | NP_612540.1 | 5 | 591 | ||
| XM_006236726.2 | XP_006236788.1 | 6 | 1258 | |||
| XM_006236727.2 | XP_006236789.1 | 7 | 1376 | |||
| XM_008763045.1 | XP_008761267.1 | 5 | 2919 | |||
| XM_008763046.1 | XP_008761268.1 | 6 | 1939 | |||
| XM_008763047.1 | XP_008761269.1 | 6 | 2356 | |||
| XM_008763048.1 | XP_008761270.1 | 5 | 2238 | |||
| GenBank mRNA | AY083159.1 | - | 4 | 415 | ||
| BC061711.1 | - | 5 | 591 | 5597943 | ||

Stambp
Genomic Map